Potassium borohydride, 98+%, Thermo Scientific Chemicals
Potassium borohydride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium borohydride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
100 g
500 g
Packaging:
View all
Plastic drum
Catalog number 196550025
Price (USD)/ Each
960.00
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
960.00
Add to cart
Potassium borohydride, 98+%, Thermo Scientific Chemicals
Catalog number196550025
Price (USD)/ Each
960.00
-
Add to cart
Chemical Identifiers
CAS13762-51-1
IUPAC Namepotassium boranuide
Molecular FormulaBH4K
InChI KeyICRGAIPBTSPUEX-UHFFFAOYSA-N
SMILES[BH4-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration>=98.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only