Thermo Scientific Chemicals

Potassium thiocyanate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 196580100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Potassium thiocyanate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 196580100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 196580100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
333-20-0
IUPAC Name
potassium cyanosulfanide
Molecular Formula
CKNS
InChI Key
ZNNZYHKDIALBAK-UHFFFAOYSA-M
SMILES
[K+].[S-]C#N
Iron (Fe)
=<2 ppm
Iodine consuming subst.
=<30 mg I2/100 g
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Titration Argentometric
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text