Potassium thiocyanate, 99+%, for analysis, Thermo Scientific Chemicals
Potassium thiocyanate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium thiocyanate, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 196582500
Price (USD)/ Each
46.65
Online exclusive
55.10 
Save 8.45 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
46.65
Online exclusive
55.10 
Save 8.45 (15%)
Add to cart
Potassium thiocyanate, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number196582500
Price (USD)/ Each
46.65
Online exclusive
55.10 
Save 8.45 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namepotassium cyanosulfanide
Molecular FormulaCKNS
InChI KeyZNNZYHKDIALBAK-UHFFFAOYSA-M
SMILES[K+].[S-]C#N
Molecular Weight (g/mol)97.18
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<2 ppm
Iodine consuming subst.=<30 mg I2/100 g
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Argentometric>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only