Chromium(VI) oxide, 99%, Thermo Scientific Chemicals
Chromium(VI) oxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium(VI) oxide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
250 g
1 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 196610025
Price (USD)/ Each
130.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
153.00 
Save 22.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
130.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
153.00 
Save 22.35 (15%)
Add to cart
Chromium(VI) oxide, 99%, Thermo Scientific Chemicals
Catalog number196610025
Price (USD)/ Each
130.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
153.00 
Save 22.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametrioxochromium
Molecular FormulaCrO3
InChI KeyWGLPBDUCMAPZCE-UHFFFAOYSA-N
SMILESO=[Cr](=O)=O
Molecular Weight (g/mol)99.99
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodometric98.5 to 101.5 %
Appearance (Color)Red to dark red to purple
Appearance (Form)Crystals or granules or flakes or prills
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only