Thermo Scientific Chemicals

Chromium(VI) oxide, 99%, Thermo Scientific Chemicals

Catalog number: 196610250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Chromium(VI) oxide, 99%, Thermo Scientific Chemicals

Catalog number: 196610250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 196610250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1333-82-0
IUPAC Name
trioxochromium
Molecular Formula
CrO3
InChI Key
WGLPBDUCMAPZCE-UHFFFAOYSA-N
SMILES
O=[Cr](=O)=O
Titration Iodometric
98.5 to 101.5 %
Appearance (Color)
Red to dark red to purple
Appearance (Form)
Crystals or granules or flakes or prills

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text