Thermo Scientific Chemicals

Sodium sulfite, 97%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 196630010
1 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Sodium sulfite, 97%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 196630010
1 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 196630010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7757-83-7
IUPAC Name
disodium heptahydrate sulfite
Molecular Formula
H14Na2O10S
InChI Key
WMJRPJZQQSSDBU-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O
Heavy metals (as Pb)
=<10 ppm
Iron (Fe)
=<10 ppm
Appearance (Form)
Crystalline powder or crystals
pH
8.5 to 10.5 (5 % solution)
Sodium chloride (NaCl)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text