Sodium sulfate, 99%, extra pure, anhydrous
Sodium sulfate, 99%, extra pure, anhydrous
Sodium sulfate, 99%, extra pure, anhydrous
Sodium sulfate, 99%, extra pure, anhydrous
Thermo Scientific Chemicals

Sodium sulfate, 99%, extra pure, anhydrous

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
25 kg
2.5 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 196640025
Price (USD)
73.50
Each
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
73.50
Each
Add to cart
Chemical Identifiers
CAS7757-82-6
IUPAC Namedisodium sulfate
Molecular FormulaNa2O4S
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Calcium (Ca)=<300 ppm
Appearance (Form)Crystalline powder
Water=<0.1 %
Iron (Fe)=<10 ppm
Magnesium (Mg)=<300 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link