Thermo Scientific Chemicals

Sodium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 196640050
5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 196640050
5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 196640050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7757-82-6
IUPAC Name
disodium sulfate
Molecular Formula
Na2O4S
InChI Key
PMZURENOXWZQFD-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]S([O-])(=O)=O
Calcium (Ca)
=<300 ppm
Appearance (Form)
Crystalline powder
Chloride (Cl)
=<3000 ppm (as NaCl)
Appearance (Color)
White
Titration after Ion exchange
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text