Sodium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
Sodium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
25 g
1 kg
2.5 kg
5 kg
Packaging:
View all
Fibre drum
Catalog number 196640251
Price (USD)/ Each
218.65
Online exclusive
243.00 
Save 24.35 (10%)
-
Add to cart
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Price (USD)/ Each
218.65
Online exclusive
243.00 
Save 24.35 (10%)
Add to cart
Sodium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
Catalog number196640251
Price (USD)/ Each
218.65
Online exclusive
243.00 
Save 24.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7757-82-6
IUPAC Namedisodium sulfate
Molecular FormulaNa2O4S
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Calcium (Ca)=<300 ppm
Water=<0.1 %
Iron (Fe)=<10 ppm
Magnesium (Mg)=<300 ppm
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only