Thermo Scientific Chemicals

Copper(II) carbonate basic, pure, 54-56% Cu, Thermo Scientific Chemicals

Catalog number: 196670025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Copper(II) carbonate basic, pure, 54-56% Cu, Thermo Scientific Chemicals

Catalog number: 196670025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 196670025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12069-69-1
IUPAC Name
dicopper(2+) dihydroxide carbonate
Molecular Formula
CH2Cu2O5
InChI Key
ZMMDPCMYTCRWFF-UHFFFAOYSA-J
SMILES
[OH-].[OH-].[Cu++].[Cu++].[O-]C([O-])=O
Assay
54 to 56 % (Cu)
Appearance (Color)
Green
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text