Hydrazine hydrate, 100% (Hydrazine, 64%)
Hydrazine hydrate, 100% (Hydrazine, 64%)
Hydrazine hydrate, 100% (Hydrazine, 64%)
Hydrazine hydrate, 100% (Hydrazine, 64%)
Hydrazine hydrate, 100% (Hydrazine, 64%)
Hydrazine hydrate, 100% (Hydrazine, 64%)
Thermo Scientific Chemicals

Hydrazine hydrate, 100% (Hydrazine, 64%)

CAS: 10217-52-4 | H4N2 | 32.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 196715000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10217-52-4
IUPAC Namehydrazine
Molecular FormulaH4N2
InChI KeyOAKJQQAXSVQMHS-UHFFFAOYSA-N
SMILESNN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Titration with HCl63.6 to 64.6 % (N2H4)
Density(20°C) 1.030 to 1.033 g/ml
Appearance (Form)Liquid
Refractive index1.4285 to 1.4315 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Share catalog number, name or link