Thermo Scientific Chemicals

Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals

Catalog number: 196731000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals

Catalog number: 196731000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 196731000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7681-11-0
IUPAC Name
potassium iodide
Molecular Formula
IK
InChI Key
NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES
[K+].[I-]
Thiosulphate (S2O3)
=<100 ppm
Heavy metals (as Pb)
=<10 ppm
Iodate (IO3)
=<4 ppm
Appearance (Color)
White
Titration Argentometric
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text