Potassium iodide, 99%, extra pure, briquettes
Potassium iodide, 99%, extra pure, briquettes
Potassium iodide, 99%, extra pure, briquettes
Potassium iodide, 99%, extra pure, briquettes
Thermo Scientific Chemicals

Potassium iodide, 99%, extra pure, briquettes

CAS: 7681-11-0 | IK | 166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 196735000
Price (USD)
200.00
Each
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
200.00
Each
Add to cart
Chemical Identifiers
CAS7681-11-0
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Thiosulphate (S2O3)=<100 ppm
Heavy metals (as Pb)=<10 ppm
Iodate (IO3)=<4 ppm
Appearance (Color)White
Titration Argentometric98.5 to 101.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link