Potassium iodate, 98%, extra pure, Thermo Scientific Chemicals
Potassium iodate, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium iodate, 98%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Glass bottle
Catalog number 196740025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namepotassium iodate
Molecular FormulaIKO3
InChI KeyJLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES[K+].[O-][I](=O)=O
Molecular Weight (g/mol)214.00
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Loss on drying=<0.5 % (105°C, 3 hrs)
Titration Iodometric>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only