Potassium chloride, 99+%, for analysis
Potassium chloride, 99+%, for analysis
Potassium chloride, 99+%, for analysis
Potassium chloride, 99+%, for analysis
Thermo Scientific Chemicals

Potassium chloride, 99+%, for analysis

CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 196770010
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric99.0 to 100.5 % (on dried substance)
Aluminium (Al)=<1 ppm
Calcium (Ca)=<50 ppm
Sodium (Na)=<700 ppm
Heavy metals (as Pb)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysis
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link