Thermo Scientific Chemicals

Potassium chloride, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 196770050
5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium chloride, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 196770050
5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 196770050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7447-40-7
IUPAC Name
potassium chloride
Molecular Formula
ClK
InChI Key
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES
[Cl-].[K+]
Titration Argentometric
99.0 to 100.5 % (on dried substance)
Aluminium (Al)
=<1 ppm
Calcium (Ca)
=<50 ppm
Sodium (Na)
=<700 ppm
Heavy metals (as Pb)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text