Thermo Scientific Chemicals

Potassium ferricyanide, 98%, pure, Thermo Scientific Chemicals

Catalog number: 196785000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Potassium ferricyanide, 98%, pure, Thermo Scientific Chemicals

Catalog number: 196785000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 196785000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13746-66-2
IUPAC Name
tripotassium hexacyanoirontris(ylium)
Molecular Formula
C6FeK3N6
InChI Key
QMTKJUMXUDIUAQ-UHFFFAOYSA-N
SMILES
[K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
Appearance (Form)
Crystalline powder or crystals
Titration Iodometric
>=97.5 %
Appearance (Color)
Orange to red

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text