Sodium hexafluorophosphate, 98.5+%, pure, Thermo Scientific Chemicals
Sodium hexafluorophosphate, 98.5+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hexafluorophosphate, 98.5+%, pure, Thermo Scientific Chemicals

Sodium hexafluorophosphate, 98.5+%, pure, Acros Organics, Quantity: 50g, Packaging: Plastic bottle, CAS: 21324-39-0, White to Gray, Grade: Pure, Melting Point: 200.0 deg.C, Molecular Formula: F6NaP, Molecular Weight: 167.95, Percent Purity: 98.5+%
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Plastic bottle
Catalog number 196820100
Price (USD)/ Each
45.65
Online exclusive
50.10 
Save 4.45 (9%)
-
Add to cart
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
45.65
Online exclusive
50.10 
Save 4.45 (9%)
Add to cart
Sodium hexafluorophosphate, 98.5+%, pure, Thermo Scientific Chemicals
Catalog number196820100
Price (USD)/ Each
45.65
Online exclusive
50.10 
Save 4.45 (9%)
-
Add to cart
Chemical Identifiers
CAS21324-39-0
IUPAC Namesodium hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6NaP
InChI KeyKMADQUOCJBLXRP-UHFFFAOYSA-N
SMILES[Na+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey
Assay>=98.50 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only