Zinc chloride, 98%, extra pure, Thermo Scientific Chemicals
Zinc chloride, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc chloride, 98%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
1 kg
10 kg
Packaging:
View all
Glass Bottle
Catalog number 196840025
Price (USD)/ Each
192.65
Online exclusive
226.00 
Save 33.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
192.65
Online exclusive
226.00 
Save 33.35 (15%)
Add to cart
Zinc chloride, 98%, extra pure, Thermo Scientific Chemicals
Catalog number196840025
Price (USD)/ Each
192.65
Online exclusive
226.00 
Save 33.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
Molecular Weight (g/mol)136.28
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Complexometric97.5 to 102.5 %
Cadmium (Cd)=<10 ppm
Copper (Cu)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only