1x1 image pixel for data collection
Zinc chloride, 98%, extra pure
Zinc chloride, 98%, extra pure
Zinc chloride, 98%, extra pure
Zinc chloride, 98%, extra pure
Thermo Scientific Chemicals

Zinc chloride, 98%, extra pure

CAS: 7646-85-7 | Cl2Zn | 136.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
5 g
1 kg
2.5 kg
Packaging:
View all
Plastic drum
Catalog number 196840100
Price (USD)
425.65
Special offer
Online exclusive
Ends: 26-Sep-2025
526.00
Save 100.35 (19%)
Each
In stock
Add to cart
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
425.65
Special offer
Online exclusive
Ends: 26-Sep-2025
526.00
Save 100.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Complexometric97.5 to 102.5 %
Cadmium (Cd)=<10 ppm
Copper (Cu)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Generating response

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection