Mercury(II) thiocyanate, 99+%
Mercury(II) thiocyanate, 99+%
Mercury(II) thiocyanate, 99+%
Mercury(II) thiocyanate, 99+%
Thermo Scientific Chemicals

Mercury(II) thiocyanate, 99+%

CAS: 592-85-8 | C4HgK2N4S4 | 511.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 196890500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS592-85-8
IUPAC Namemercury(2+) dipotassium tetrakis(cyanosulfanide)
Molecular FormulaC4HgK2N4S4
InChI KeyHWXOCCNUUYRBKP-UHFFFAOYSA-J
SMILES[K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Iron (Fe)=<20 ppm
Titration Complexometric>=99.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0467030Certificate of AnalysisJan 31, 2025196890500, 196892500
A0459132Certificate of AnalysisJan 17, 2024196890500, 196892500
A0458102Certificate of AnalysisNov 27, 2023196890500, 196892500
A0449335Certificate of AnalysisMay 02, 2023196892500, 196890500
A0449335Certificate of AnalysisFeb 01, 2023196890500, 196892500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection