Calcium oxide, 96%, extra pure, Thermo Scientific Chemicals
Calcium oxide, 96%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium oxide, 96%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
View all
Plastic bottle
Catalog number 196910025
Price (USD)/ Each
211.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
249.00 
Save 37.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
211.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
249.00 
Save 37.35 (15%)
Add to cart
Calcium oxide, 96%, extra pure, Thermo Scientific Chemicals
Catalog number196910025
Price (USD)/ Each
211.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
249.00 
Save 37.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1305-78-8
IUPAC Namecalcium oxidandiide
Molecular FormulaCaO
InChI KeyBRPQOXSCLDDYGP-UHFFFAOYSA-N
SMILES[O--].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Titration Complexometric>=95 %
Insoluble matter=<0.5 % (in HCl)
Appearance (Form)Powder
Loss on ignition=<3.5 % (800°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only