Thermo Scientific Chemicals

Polyphosphoric acid, pure, >84% phosphate (as P2O5), Thermo Scientific Chemicals

Catalog number: 196950010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Polyphosphoric acid, pure, >84% phosphate (as P2O5), Thermo Scientific Chemicals

Catalog number: 196950010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 196950010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
8017-16-1
IUPAC Name
Polyphosphoric acid
Molecular Formula
(HO3P)n
InChI Key
NBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILES
[H]OP(O)(O)=O
Appearance (Form)
Clear viscous liquid
Titration with NaOH
>=82 % (as P2O5)
Color scale
=<50 APHA
Appearance
May darken on storage

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text