Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Polyphosphoric acid, pure, > 84% phosphate (as P2O5)
Thermo Scientific Chemicals

Polyphosphoric acid, pure, > 84% phosphate (as P2O5)

CAS: 8017-16-1 | (HO3P)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Plastic bottle
Catalog number 196950025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS8017-16-1
IUPAC NamePolyphosphoric acid
Molecular Formula(HO3P)n
InChI KeyNBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILES[H]OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear viscous liquid
Titration with NaOH>=82 % (as P2O5)
Color scale=<50 APHA
AppearanceMay darken on storage
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only