Aluminum acetylacetonate, 97%
Aluminum acetylacetonate, 97%
Aluminum acetylacetonate, 97%
Aluminum acetylacetonate, 97%
Thermo Scientific Chemicals

Aluminum acetylacetonate, 97%

CAS: 13963-57-0 | C15H21AlO6 | 324.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 196975000
Price (USD)
92.65
Special offer
Online exclusive
109.00
Save 16.35 (15%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
109.00
Save 16.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS13963-57-0
IUPAC Namealuminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21AlO6
InChI KeyKILURZWTCGSYRE-LNTINUHCSA-K
SMILES[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Complexometric>=96.0 %
Appearance (Color)White to cream
Melting point190°C to 195°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection