Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals
Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 196980250
Price (USD)/ Each
32.65
Online exclusive
35.80 
Save 3.15 (9%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
32.65
Online exclusive
35.80 
Save 3.15 (9%)
Add to cart
Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals
Catalog number196980250
Price (USD)/ Each
32.65
Online exclusive
35.80 
Save 3.15 (9%)
-
Add to cart
Chemical Identifiers
CAS7772-99-8
IUPAC Nameλ²-tin(2+) dichloride
Molecular FormulaCl2Sn
InChI KeyAXZWODMDQAVCJE-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Iron (Fe)=<50 ppm
Appearance (Form)Crystalline powder or flakes
Lead (Pb)=<75 ppm
Titration Iodimetric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only