Thermo Scientific Chemicals

Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals

Catalog number: 196981000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals

Catalog number: 196981000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 196981000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7772-99-8
IUPAC Name
λ²-tin(2+) dichloride
Molecular Formula
Cl2Sn
InChI Key
AXZWODMDQAVCJE-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Sn++]
Appearance (Form)
Crystalline powder or flakes
Appearance (Color)
White to off-white
Iron (Fe)
=<50 ppm
Lead (Pb)
=<75 ppm
Titration Iodimetric
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text