Thermo Scientific Chemicals

Allyltrimethylsilane, 99%, Thermo Scientific Chemicals

Catalog number: 196990100
10 mL, Glass bottle, Each
Thermo Scientific Chemicals

Allyltrimethylsilane, 99%, Thermo Scientific Chemicals

Catalog number: 196990100
10 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 196990100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
762-72-1
IUPAC Name
trimethyl(prop-2-en-1-yl)silane
Molecular Formula
C6H14Si
InChI Key
HYWCXWRMUZYRPH-UHFFFAOYSA-N
SMILES
C[Si](C)(C)CC=C
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4060 to 1.4080 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text