Allyltrimethylsilane, 99%, Thermo Scientific Chemicals
Allyltrimethylsilane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Allyltrimethylsilane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
250 mL
Packaging:
Glass bottle
Catalog number 196990500
Price (USD)/ Each
173.65
Online exclusive
193.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
173.65
Online exclusive
193.00 
Save 19.35 (10%)
Add to cart
Allyltrimethylsilane, 99%, Thermo Scientific Chemicals
Catalog number196990500
Price (USD)/ Each
173.65
Online exclusive
193.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Nametrimethyl(prop-2-en-1-yl)silane
Molecular FormulaC6H14Si
InChI KeyHYWCXWRMUZYRPH-UHFFFAOYSA-N
SMILESC[Si](C)(C)CC=C
Molecular Weight (g/mol)114.26
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4060 to 1.4080 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only