Thermo Scientific Chemicals

Cadmium chloride hemipentahydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 197022500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Cadmium chloride hemipentahydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 197022500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 197022500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7790-78-5
IUPAC Name
cadmium(2+) dichloride
Molecular Formula
CdCl2
InChI Key
YKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Cd++]
Sulfate (SO4)
=<50 ppm
Lead (Pb)
=<50 ppm
Appearance (Form)
Crystalline powder or crystals and/or chunks
Insoluble matter
=<0.005 % (in water)
Ammonium (NH4)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text