Sodium amide, 95%, pure
Sodium amide, 95%, pure
Sodium amide, 95%, pure
Sodium amide, 95%, pure
Thermo Scientific Chemicals

Sodium amide, 95%, pure

CAS: 7782-92-5 | H2NNa | 39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 197051000
Price (USD)
43.65
Online Exclusive
48.80
Save 5.15 (11%)
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
43.65
Online Exclusive
48.80
Save 5.15 (11%)
Each
Add to cart
Chemical Identifiers
CAS7782-92-5
IUPAC Namesodium azanide
Molecular FormulaH2NNa
InChI KeyODZPKZBBUMBTMG-UHFFFAOYSA-N
SMILES[NH2-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to greyish-white
Free SodiumPasses test (A small amount dropped into water,
Free Sodiumdoes not catch fire)
Appearance (Form)Powder or irregular granules or flakes
Peroxides=<0.1 % (as NaNH2O2)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link