Thermo Scientific Chemicals

Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 197060500
50 g, Plastic bottle, Each
Thermo Scientific Chemicals

Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 197060500
50 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 197060500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7786-30-3
IUPAC Name
magnesium(2+) dichloride
Molecular Formula
Cl2Mg
InChI Key
TWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES
[Mg++].[Cl-].[Cl-]
Total trace metal impurities
=<0.1 %
Appearance (Form)
Powder to beads
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text