Thermo Scientific Chemicals

Molybdenum(V) chloride, 95%, Thermo Scientific Chemicals

Catalog number: 197080100
10 g, Glass Bottle, Each
Thermo Scientific Chemicals

Molybdenum(V) chloride, 95%, Thermo Scientific Chemicals

Catalog number: 197080100
10 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 197080100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10241-05-1
IUPAC Name
molybdenum(5+) pentachloride
Molecular Formula
Cl5Mo
InChI Key
GICWIDZXWJGTCI-UHFFFAOYSA-I
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5]
Appearance (Color)
Blue to green-black to black
Titration Argentometric
>=94 %
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text