Molybdenum(V) chloride, 95%, Thermo Scientific Chemicals
Molybdenum(V) chloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum(V) chloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Packaging:
View all
Glass bottle
Catalog number 197081000
Price (USD)/ Each
189.65
Online exclusive
211.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
189.65
Online exclusive
211.00 
Save 21.35 (10%)
Add to cart
Molybdenum(V) chloride, 95%, Thermo Scientific Chemicals
Catalog number197081000
Price (USD)/ Each
189.65
Online exclusive
211.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10241-05-1
IUPAC Namemolybdenum(5+) pentachloride
Molecular FormulaCl5Mo
InChI KeyGICWIDZXWJGTCI-UHFFFAOYSA-I
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue to green-black to black
Titration Argentometric>=94 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only