Silver tetrafluoroborate, 99%, Thermo Scientific Chemicals
Silver tetrafluoroborate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver tetrafluoroborate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
10 g
Packaging:
Plastic bottle
Catalog number 197090500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS14104-20-2
IUPAC Namesilver(1+) tetrafluoroboranuide
Molecular FormulaAgBF4
InChI KeyCCAVYRRHZLAMDJ-UHFFFAOYSA-N
SMILES[Ag+].F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Assay>=54.58 % (Ag)
Assay>=43.92 % (BF4)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only