Sodium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals
Sodium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 197120051
Price (USD)/ Each
851.65
Online exclusive
946.00 
Save 94.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
851.65
Online exclusive
946.00 
Save 94.35 (10%)
Add to cart
Sodium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number197120051
Price (USD)/ Each
851.65
Online exclusive
946.00 
Save 94.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7601-89-0
IUPAC Namesodium perchlorate
Molecular FormulaClNaO4
InChI KeyBAZAXWOYCMUHIX-UHFFFAOYSA-M
SMILES[Na+].[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Calcium (Ca)=<200 ppm
Iron (Fe)=<5 ppm
Potassium (K)=<500 ppm
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only