1x1 image pixel for data collection
Sodium perchlorate, 99+%, for analysis
Sodium perchlorate, 99+%, for analysis
Sodium perchlorate, 99+%, for analysis
Sodium perchlorate, 99+%, for analysis
Thermo Scientific Chemicals

Sodium perchlorate, 99+%, for analysis

CAS: 7601-89-0 | ClNaO4 | 122.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 197122500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7601-89-0
IUPAC Namesodium perchlorate
Molecular FormulaClNaO4
InChI KeyBAZAXWOYCMUHIX-UHFFFAOYSA-M
SMILES[Na+].[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<20 ppm
Heavy metals (as Pb)=<5 ppm
Chloride (Cl)=<30 ppm
pH6.0 to 8.0 (5 % in water)
Insoluble matter=<0.005 % (in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection