Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh, Thermo Scientific Chemicals
Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 197140100
Price (USD)/ Each
112.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
132.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
112.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
132.00 
Save 19.35 (15%)
Add to cart
Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh, Thermo Scientific Chemicals
Catalog number197140100
Price (USD)/ Each
112.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
132.00 
Save 19.35 (15%)
-
Add to cart
Chemical Identifiers
CAS12055-62-8
IUPAC Namediholmium(3+) trioxidandiide
Molecular FormulaHo2O3
InChI KeyOWCYYNSBGXMRQN-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Ho+3].[Ho+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on ignition=<1.0 % (1000°C)
Appearance (Form)Powder
Particle size=<325 mesh
Appearance (Color)Yellow to beige to light orange
Assay>=99.85 % (Ho2O3) TREO base
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only