Germanium tetrachloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Germanium tetrachloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Germanium tetrachloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 197151000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10038-98-9
IUPAC Namegermanium tetrachloride
Molecular FormulaCl4GeH8
InChI KeyVDHBHZZBEGQJRK-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.[GeH4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Total trace metal impurities=<0.01 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only