Molecular sieves 3A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 3A, 8 to 12 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molecular sieves 3A, 8 to 12 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
25 kg
Catalog number 197255000
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
Add to cart
Molecular sieves 3A, 8 to 12 mesh, Thermo Scientific Chemicals
Catalog number197255000
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS308080-99-1
IUPAC NameMolecular sieves 3A
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light grey to beige
Appearance (Form)Beads
Bulk density680 to 800 g/l
Residual water=<1.5 % (550°C, 2h)
Water adsorption>=21 % (50% rH, 25°C, 1bar)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only