Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
25 kg
Packaging:
View all
Glass bottle
Catalog number 197275000
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
Add to cart
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Catalog number197275000
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS70955-01-0
IUPAC NameMolecular sieves
Synonym2-[2-acetamido-3-(3-fluorophenyl)propanamido]-5-methoxy-4-(methoxycarbonyl)-3-methyl-5-oxopentanoic acid
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light grey to beige
Appearance (Form)Beads
Bulk density680 to 800 g/l
Residual water=<1.5 % (550°C, 2h)
Water adsorption>=21 % (50%rH, 25°C, 1bar)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only