Molecular sieves 5A, 4 to 8 mesh, Thermo Scientific Chemicals
Molecular sieves 5A, 4 to 8 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molecular sieves 5A, 4 to 8 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
View all
Plastic Bottle
Catalog number 197280025
Price (USD)/ Each
204.65
Online exclusive
241.00 
Save 36.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
204.65
Online exclusive
241.00 
Save 36.35 (15%)
Add to cart
Molecular sieves 5A, 4 to 8 mesh, Thermo Scientific Chemicals
Catalog number197280025
Price (USD)/ Each
204.65
Online exclusive
241.00 
Save 36.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC NameMolecular sieves 5A
MDL NumberMFCD00147627
CAS69912-79-4
SpecificationsSpecification SheetSpecification Sheet
Bulk density>=680 g/l
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only