Molecular sieves 5A, 4 to 8 mesh
Molecular sieves 5A, 4 to 8 mesh
Molecular sieves 5A, 4 to 8 mesh
Molecular sieves 5A, 4 to 8 mesh
Thermo Scientific Chemicals

Molecular sieves 5A, 4 to 8 mesh

CAS: 69912-79-4 | AlNaO6Si2 | 202.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 197285000
Price (USD)
66.65
Special offer
Online exclusive
Ends: 26-Sep-2025
82.50
Save 15.85 (19%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
66.65
Special offer
Online exclusive
Ends: 26-Sep-2025
82.50
Save 15.85 (19%)
Each
Add to cart
Chemical Identifiers
CAS69912-79-4
IUPAC NameMolecular sieves 5A
MDL NumberMFCD00147627
SpecificationsSpecification SheetSpecification Sheet
Bulk density>=680 g/l
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473754Certificate of AnalysisJun 25, 2025197285000, 197280025
A0472085Certificate of AnalysisMay 17, 2025197285000, 197280025
A0469842Certificate of AnalysisFeb 13, 2025197285000, 197280025
A0467593Certificate of AnalysisNov 11, 2024197285000, 197280025
A0465703Certificate of AnalysisAug 29, 2024197285000, 197280025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection