Molecular sieves 5A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 5A, 8 to 12 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molecular sieves 5A, 8 to 12 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
Packaging:
View all
Plastic drum
Catalog number 197290050
Price (USD)/ Each
332.65
Online exclusive
391.00 
Save 58.35 (15%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
332.65
Online exclusive
391.00 
Save 58.35 (15%)
Add to cart
Molecular sieves 5A, 8 to 12 mesh, Thermo Scientific Chemicals
Catalog number197290050
Price (USD)/ Each
332.65
Online exclusive
391.00 
Save 58.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC NameMolecular sieves 5A
MDL NumberMFCD00147627
CAS69912-79-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light grey to beige
Appearance (Form)Beads
Bulk density680 to 800 g/l
Residual water=<1.5 % (550°C, 2h)
Water adsorption>=21 % (50%rH, 25°C, 1 bar)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only