Molecular sieves 13X, 4 to 8 mesh, Thermo Scientific Chemicals
Molecular sieves 13X, 4 to 8 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molecular sieves 13X, 4 to 8 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
Packaging:
View all
Plastic drum
Catalog number 197300050
Price (USD)/ Each
322.65
Online exclusive
358.00 
Save 35.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
322.65
Online exclusive
358.00 
Save 35.35 (10%)
Add to cart
Molecular sieves 13X, 4 to 8 mesh, Thermo Scientific Chemicals
Catalog number197300050
Price (USD)/ Each
322.65
Online exclusive
358.00 
Save 35.35 (10%)
-
Add to cart
Chemical Identifiers
CAS63231-69-6
IUPAC NameMolecular sieves 13X
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light grey to beige
Appearance (Form)Beads
Bulk density>=600 g/l
Residual water=<1.5 % (550°C, 2h)
Water adsorption>=26 % (50%rH, 25°C, 1bar)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only