S-Methyl methanethiolsulfonate, 97%, Thermo Scientific Chemicals
S-Methyl methanethiolsulfonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

S-Methyl methanethiolsulfonate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 197390010
Price (USD)/ Each
42.50
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
42.50
Add to cart
S-Methyl methanethiolsulfonate, 97%, Thermo Scientific Chemicals
Catalog number197390010
Price (USD)/ Each
42.50
-
Add to cart
Chemical Identifiers
IUPAC Name(methanesulfonylsulfanyl)methane
Molecular FormulaC2H6O2S2
InChI KeyXYONNSVDNIRXKZ-UHFFFAOYSA-N
SMILESCSS(C)(=O)=O
Molecular Weight (g/mol)126.19
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear yellow to orange
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only