Thermo Scientific Chemicals

S-Methyl methanethiolsulfonate, 97%, Thermo Scientific Chemicals

Catalog number: 197390250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

S-Methyl methanethiolsulfonate, 97%, Thermo Scientific Chemicals

Catalog number: 197390250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 197390250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2949-92-0
IUPAC Name
(methanesulfonylsulfanyl)methane
Molecular Formula
C2H6O2S2
InChI Key
XYONNSVDNIRXKZ-UHFFFAOYSA-N
SMILES
CSS(C)(=O)=O
Infrared spectrum
Conforms
Appearance (Color)
Clear yellow to orange
Appearance (Form)
Liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text