1x1 image pixel for data collection
Copper(I) oxide, 97%
Copper(I) oxide, 97%
Copper(I) oxide, 97%
Copper(I) oxide, 97%
Thermo Scientific Chemicals

Copper(I) oxide, 97%

CAS: 1317-39-1 | Cu2O | 143.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 197420250
Price (USD)
51.65
Special offer
Online exclusive
Ends: 26-Sep-2025
63.60
Save 11.95 (19%)
Each
In stock
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 26-Sep-2025
63.60
Save 11.95 (19%)
Each
Add to cart
Chemical Identifiers
CAS1317-39-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Nickel (Ni)=<100 ppm
Silver (Ag)=<50 ppm
Titration Complexometric>=96.0 %
Iron (Fe)=<500 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473604Certificate of AnalysisJun 26, 2025197425000, 197420025, 197420250
A0471479Certificate of AnalysisMay 17, 2025197425000, 197420025, 197420250
A0468186Certificate of AnalysisJan 31, 2025197425000, 197420025, 197420250
A0466018Certificate of AnalysisSep 19, 2024197425000, 197420025, 197420250
A0454403Certificate of AnalysisAug 25, 2023197425000, 197420025, 197420250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection