Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized, Thermo Scientific Chemicals
Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
1 mL
25 mL
Packaging:
Glass bottle
Catalog number 197450050
Price (USD)/ Each
124.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
147.00 
Save 22.35 (15%)
-
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
124.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
147.00 
Save 22.35 (15%)
Add to cart
Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized, Thermo Scientific Chemicals
Catalog number197450050
Price (USD)/ Each
124.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
147.00 
Save 22.35 (15%)
-
Add to cart
Chemical Identifiers
CAS20816-12-0, 75-65-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Appearance (Form)Fused mass or solution
Strength>=2.5 wt%
Stabilizertert-Butyl hydroperoxide
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only