Thermo Scientific Chemicals

Urea, 98%, extra pure, pearls, Thermo Scientific Chemicals

Catalog number: 197460100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Urea, 98%, extra pure, pearls, Thermo Scientific Chemicals

Catalog number: 197460100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 197460100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-13-6
IUPAC Name
urea
Molecular Formula
CH4N2O
InChI Key
XSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILES
NC(N)=O
Water
=<0.4 %
Impurity
=<1.2 % (Biuret)
Appearance (Color)
White
Appearance (Form)
Pearls
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text